Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HELLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289067.1 | NP_001275996.1 | 22 | 2693 | ||
NM_001289068.1 | NP_001275997.1 | 21 | 2510 | |||
NM_001289069.1 | NP_001275998.1 | 21 | 2459 | |||
NM_001289070.1 | NP_001275999.1 | 19 | 2261 | |||
NM_001289071.1 | NP_001276000.1 | 22 | 2599 | |||
NM_001289072.1 | NP_001276001.1 | 19 | 2165 | |||
NM_001289073.1 | NP_001276002.1 | 20 | 2490 | |||
NM_001289074.1 | NP_001276003.1 | 22 | 2474 | |||
NM_001289075.1 | NP_001276004.1 | 20 | 2358 | |||
NM_018063.4 | NP_060533.2 | 21 | 2555 | |||
GenBank mRNA | AB074174.1 | - | 9 | 1003 | ||
AB102716.1 | - | 22 | 2444 | |||
AB102717.1 | - | 20 | 2335 | |||
AB102718.1 | - | 21 | 2304 | |||
AB102719.1 | - | 19 | 2010 | |||
AB102720.1 | - | 20 | 2203 | |||
AB102721.1 | - | 19 | 2106 | |||
AB102722.1 | - | 22 | 2319 | |||
AB102723.1 | - | 22 | 2538 | |||
AB113248.1 | - | 21 | 2313 | |||
AB113249.1 | - | 20 | 2478 | |||
AF155827.1 | - | 21 | 2505 | |||
AK001201.1 | - | 7 | 713 | |||
AK314485.1 | - | 21 | 2544 | |||
AY007108.1 | - | 9 | 1012 | |||
BC015477.1 | - | 8 | 838 | |||
BC029381.1 | - | 9 | 943 | |||
BC030963.1 | - | 10 | 1158 | |||
BC031004.1 | - | 10 | 1158 | |||
BX538033.1 | - | 21 | 2494 |