Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110831.1 | NP_001104301.1 | 4, 5 | 601 | ||
NM_016878.4 | NP_058574.3 | 4, 5 | 425 | |||
GenBank mRNA | AF005051.1 | - | 3, 4 | 535 | ||
AK038646.1 | - | 601 | ||||
AK052001.1 | - | 403 | ||||
AK160084.1 | - | 4, 5 | 362 | |||
AK182360.1 | - | 1, 2 | 84 | |||
BC092232.1 | - | 4, 5 | 353 | 30551654 |