Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010073.2 | NP_034203.1 | 4 | 377 | ||
GenBank mRNA | AK002468.1 | - | 4 | 361 | ||
AK004241.1 | - | 4 | 352 | |||
BC008256.1 | - | 4 | 377 | 2648752 |