Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007877.1 | NP_031903.1 | 7 | 1189 | ||
GenBank mRNA | BC119286.1 | - | 7 | 1210 | 8734035 | |
BC137819.1 | - | 7 | 1210 | 8860838 | ||
X67274.1 | - | 7 | 1189 |
Set Membership: |
![]() |