Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Opn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010098.3 | NP_034228.1 | 3 | 971 | ||
GenBank mRNA | AK043135.1 | - | 3 | 972 | ||
AK132822.1 | - | 3 | 908 |
Set Membership: |
![]() |