Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177937.1 | NP_001171408.1 | 8 | 1247 | ||
NM_001177938.1 | NP_001171409.1 | 8 | 1244 | |||
NM_001177939.1 | NP_001171410.1 | 8 | 1238 | |||
NM_001177940.1 | NP_001171411.1 | 7 | 1229 | |||
NM_001177941.1 | NP_001171412.1 | 7 | 1214 | |||
NM_001177942.1 | NP_001171413.1 | 8 | 1211 | |||
NM_001177943.1 | NP_001171414.1 | 9 | 1403 | |||
NM_001177944.1 | NP_001171415.1 | 9 | 1386 | |||
NM_010099.2 | NP_034229.1 | 8 | 1253 | |||
GenBank mRNA | AF004435.1 | - | 8 | 1177 | ||
AF016628.1 | - | 8 | 1253 | |||
AF016629.1 | - | 8 | 1253 | |||
AJ243657.1 | - | 8 | 1234 | |||
AJ243658.1 | - | 8 | 1186 | |||
AK137551.1 | - | 8 | 1162 | |||
BC125387.1 | - | 8 | 1144 | 40129902 | ||
BC144790.1 | - | 8 | 1135 | 9053338 | ||
BC144791.1 | - | 8 | 1138 | 9053339 | ||
DQ168876.1 | - | 8 | 1253 | |||
DQ168877.1 | - | 8 | 1244 | |||
DQ168878.1 | - | 7 | 1229 | |||
DQ168879.1 | - | 7 | 1214 | |||
DQ168880.1 | - | 9 | 1403 | |||
DQ168881.1 | - | 9 | 1386 |