Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HGFAC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297439.1 | NP_001284368.1 | 6 | 682 | ||
NM_001528.3 | NP_001519.1 | 6 | 682 | |||
GenBank mRNA | AK314099.1 | - | 6 | 640 | ||
D14012.1 | - | 6 | 616 |