Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013508.2 | NP_038536.2 | 3 | 978 | ||
GenBank mRNA | AK052546.1 | - | 3 | 655 | ||
AK133847.1 | - | 3 | 674 | |||
BC005686.1 | - | 3 | 960 | 3155850 | ||
BC053402.1 | - | 3 | 729 | 30060743 | ||
BC055735.1 | - | 3 | 751 | 6808738 | ||
L19953.1 | - | 3 | 869 | |||
S82864.1 | - | 3 | 661 | |||
Z32815.1 | - | 3 | 862 |