Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010130.4 | NP_034260.1 | 16 | 1978 | ||
GenBank mRNA | AK133202.1 | - | 16 | 1954 | ||
AK149898.1 | - | 16 | 1963 | |||
AK151303.1 | - | 16 | 1950 | |||
AK151682.1 | - | 16 | 1970 | |||
AK155862.1 | - | 16 | 1929 | |||
BC075688.1 | - | 16 | 1950 | 30609706 | ||
DQ167573.1 | - | 2 | 162 | |||
U66888.1 | - | 16 | 1965 | |||
X93328.1 | - | 16 | 1974 |
Set Membership: |
![]() |