Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006664.3 | NP_001006665.1 | 16 | 2581 | ||
NM_001291120.1 | NP_001278049.1 | 17 | 2548 | |||
NM_001291121.1 | NP_001278050.1 | 16 | 2512 | |||
NM_001291122.1 | NP_001278051.1 | 15 | 2104 | |||
NM_001291123.1 | NP_001278052.1 | 15 | 2101 | |||
NM_013510.4 | NP_038538.1 | 16 | 2499 | |||
GenBank mRNA | AB370262.1 | - | 15 | 2094 | ||
AF061283.1 | - | 16 | 2313 | |||
AK122251.1 | - | 16 | 2179 | |||
AK147272.1 | - | 15 | 1963 | |||
AK147611.1 | - | 17 | 2550 | |||
AK154704.1 | - | 15 | 1923 | |||
BC034751.1 | - | 16 | 2309 | 4954660 |