Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252193.1 | NP_001239122.1 | 4 | 801 | ||
NM_025275.4 | NP_079551.3 | 5 | 620 | |||
GenBank mRNA | AJ879914.1 | - | 2 | 317 | ||
AK004062.1 | - | 5 | 581 | |||
AK075748.1 | - | 5 | 560 | |||
AK143908.1 | - | 5 | 585 | |||
AK195110.1 | - | 2 | 55 | |||
BC025087.1 | - | 4 | 795 | 2649126 |