Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK149372.1 | - | 8 | 1002 | ||
BC125617.1 | - | 8 | 1000 | |||
BC132393.1 | - | 8 | 1000 | |||
M23109.1 | - | 8 | 965 | |||
M26236.1 | - | 3 | 464 |