Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080387.2 | NP_001073856.1 | 2, 3 | 319 | ||
NM_001284195.1 | NP_001271124.1 | 2, 3 | 319 | |||
NM_001284196.1 | NP_001271125.1 | 2, 3 | 319 | |||
NM_010178.3 | NP_034308.1 | 2, 3 | 319 | |||
GenBank mRNA | AB015894.1 | - | 2, 3 | 230 | ||
AB015895.1 | - | 2, 3 | 227 | |||
AF042383.1 | - | 2, 3 | 231 | |||
AF060490.1 | - | 2, 3 | 254 | |||
AK014345.1 | - | 2, 3 | 258 | |||
AK134076.1 | - | 2, 3 | 289 | |||
AK134289.1 | - | 2, 3 | 289 | |||
AK135316.1 | - | 2, 3 | 264 | |||
AK149716.1 | - | 2, 3 | 246 | |||
AK150072.1 | - | 2, 3 | 246 | |||
AK150734.1 | - | 2, 3 | 253 | |||
AK151567.1 | - | 2, 3 | 258 | |||
AK168524.1 | - | 2, 3 | 277 | |||
AK168558.1 | - | 2, 3 | 277 | |||
AK168571.1 | - | 2, 3 | 242 | |||
AK169071.1 | - | 2, 3 | 246 | |||
BC006670.1 | - | 2, 3 | 271 | |||
BC037591.1 | - | 2, 3 | 240 | 5004611 | ||
BC043060.1 | - | 2, 3 | 643 | 6419883 | ||
BC083082.1 | - | 2, 3 | 231 | 6511807 | ||
BY244789.1 | - | 2, 3 | 319 | |||
BY717856.1 | - | 2, 3 | 289 | |||
CF914989.1 | - | 2, 3 | 235 |