Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgrt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010189.3 | NP_034319.2 | 6 | 1300 | ||
GenBank mRNA | AK146933.1 | - | 6 | 1265 | ||
BC003786.1 | - | 6 | 1235 | 3485649 | ||
D37874.1 | - | 6 | 1233 | |||
L17022.1 | - | 5 | 939 |
Gene Symbol: Mup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC093515.1 | - | 2497 |