Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgrt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010189.3 | NP_034319.2 | 3, 4 | 705 | ||
GenBank mRNA | AK146933.1 | - | 3, 4 | 670 | ||
AK152879.1 | - | 3, 4 | 641 | |||
D37874.1 | - | 3, 4 | 638 | |||
L17022.1 | - | 2, 3 | 344 |