Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010197.3 | NP_034327.1 | 3 | 397 | ||
GenBank mRNA | AK035330.1 | - | 3 | 398 | ||
AK142163.1 | - | 4 | 474 | |||
BC027001.1 | - | 3 | 386 | |||
BC037601.1 | - | 3 | 416 | 5137246 | ||
M30641.1 | - | 2 | 210 | |||
U67610.1 | - | 3 | 363 |