Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008006.2 | NP_032032.1 | 3 | 515 | ||
GenBank mRNA | AF065903.1 | - | 3 | 318 | ||
AF065904.1 | - | 3 | 318 | |||
AF065905.1 | - | 3 | 318 | |||
AY027548.1 | - | 3 | 233 | |||
AY027549.1 | - | 3 | 338 | |||
AY027550.1 | - | 1 | 110 | |||
AY027551.1 | - | 3 | 315 | |||
AY027557.1 | - | 3 | 388 | |||
AY027558.1 | - | 3 | 379 | |||
AY027559.1 | - | 3 | 242 | |||
CJ098597.1 | - | 3 | 518 | |||
M30644.1 | - | 3 | 318 |