Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fgf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010202.5 | NP_034332.2 | 2, 3 | 546 | ||
GenBank mRNA | AK082814.1 | - | 2, 3 | 547 | ||
BC104312.1 | - | 2, 3 | 427 | 40044462 | ||
BC104313.1 | - | 2, 3 | 427 | 40044466 | ||
M30642.1 | - | 2, 3 | 427 |