Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fgf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277268.1 | NP_001264197.1 | 2 | 590 | ||
NM_010203.5 | NP_034333.1 | 3 | 694 | |||
GenBank mRNA | AB016516.1 | - | 2 | 362 | ||
AK028694.1 | - | 3 | 551 | |||
AK028894.1 | - | 3 | 696 | |||
BC071227.1 | - | 3 | 667 | 30443152 | ||
M30643.1 | - | 3 | 466 |