Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fgl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008013.4 | NP_032039.2 | 2 | 1085 | ||
GenBank mRNA | AK009500.1 | - | 2 | 1117 | ||
AK036402.1 | - | 2 | 1056 | |||
BC028893.1 | - | 2 | 1085 | 4189071 |