Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fosl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008037.4 | NP_032063.2 | 4 | 1553 | ||
GenBank mRNA | AK089371.1 | - | 2179 | |||
AK145822.1 | - | 3 | 1524 | |||
BC065131.1 | - | 4 | 1491 | 30536144 |