Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002126.4 | NP_002117.1 | 3 | 1015 | ||
XM_005257269.2 | XP_005257326.1 | 3 | 995 | |||
XM_011524705.1 | XP_011523007.1 | 3 | 469 | |||
XM_017024556.1 | XP_016880045.1 | 3 | 469 | |||
GenBank mRNA | AK074289.1 | - | 2 | 479 | ||
AK290043.1 | - | 3 | 452 | |||
AK295726.1 | - | 2 | 507 | |||
AK315079.1 | - | 3 | 653 | |||
BC036093.1 | - | 3 | 969 | 5288156 | ||
CR541672.1 | - | 3 | 490 | |||
DA637609.1 | - | 2 | 534 | |||
M95585.1 | - | 3 | 812 | |||
X68985.1 | - | 3 | 588 |