Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cidec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301295.1 | NP_001288224.1 | 4 | 291 | ||
NM_178373.3 | NP_848460.1 | 4 | 275 | |||
GenBank mRNA | AK080133.1 | - | 4 | 277 | ||
BC099676.1 | - | 3 | 247 | 5043221 | ||
M61737.1 | - | 4 | 291 |