Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204973.1 | NP_001191902.1 | 11 | 3738 | ||
NM_010238.3 | NP_034368.2 | 11 | 3557 | |||
GenBank mRNA | AB212273.1 | - | 9 | 1892 | ||
AF045462.1 | - | 10 | 2703 | |||
AK158970.1 | - | 11 | 3558 | |||
AK168525.1 | - | 11 | 3429 | |||
AK220444.1 | - | 10 | 2705 | |||
BC046809.1 | - | 11 | 2419 | |||
BC138655.1 | - | 10 | 2331 | 8861675 | ||
BC138656.1 | - | 10 | 2331 | 8861676 | ||
KJ650222.1 | - | 10 | 2023 |