Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6pc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008061.3 | NP_032087.2 | 4 | 588 | ||
GenBank mRNA | AK050279.1 | - | 4 | 603 | ||
AK052656.1 | - | 4 | 603 | |||
BC013448.1 | - | 4 | 588 | 4237115 | ||
U00445.1 | - | 4 | 603 |