Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008070.3 | NP_032096.1 | 4 | 617 | ||
GenBank mRNA | AK034069.1 | - | 3 | 232 | ||
AK090279.1 | - | 4 | 618 | |||
BC145973.1 | - | 4 | 591 | 40131276 | ||
BC145975.1 | - | 4 | 591 | 40131278 | ||
GU086167.1 | - | 3 | 190 | |||
U14419.1 | - | 3 | 281 |