Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010252.4 | NP_034382.2 | 4, 5 | 942 | ||
GenBank mRNA | AK032128.1 | - | 4, 5 | 627 | ||
AK034085.1 | - | 4, 5 | 530 | |||
AK034332.1 | - | 4, 5 | 943 | |||
AK038468.1 | - | 4, 5 | 679 | |||
AK039068.1 | - | 4, 5 | 640 | |||
AK047752.1 | - | 4, 5 | 692 | |||
AK049956.1 | - | 4, 5 | 680 | |||
AK140399.1 | - | 4, 5 | 692 | |||
AK162884.1 | - | 5, 6 | 625 | |||
AK163541.1 | - | 4, 5 | 650 | |||
BC099939.1 | - | 4, 5 | 651 | 30090762 | ||
X55272.1 | - | 4, 5 | 725 |