Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008076.3 | NP_032102.2 | 4 | 509 | ||
GenBank mRNA | AF024621.1 | - | 4 | 622 | ||
AK132642.1 | - | 4 | 337 | |||
AK160446.1 | - | 2 | 176 | |||
BC057957.1 | - | 4 | 494 | 5357713 |