Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319077.1 | NP_001306006.1 | 6 | 891 | ||
NM_001319078.1 | NP_001306007.1 | 5 | 812 | |||
NM_001319079.1 | NP_001306008.1 | 6 | 924 | |||
NM_001319080.1 | NP_001306009.1 | 6 | 923 | |||
NM_001319081.1 | NP_001306010.1 | 5 | 810 | |||
NM_001319082.1 | NP_001306011.1 | 6 | 946 | |||
NM_002131.3 | NP_002122.1 | 6 | 893 | |||
NM_145899.2 | NP_665906.1 | 6 | 926 | |||
NM_145901.2 | NP_665908.1 | 5 | 1093 | |||
NM_145902.2 | NP_665909.1 | 5 | 1060 | |||
NM_145903.2 | NP_665910.1 | 5 | 779 | |||
NM_145905.2 | NP_665912.1 | 5 | 782 | |||
GenBank mRNA | AF176039.1 | - | 4 | 598 | ||
AK096863.1 | - | 7 | 833 | |||
AK301434.1 | - | 5 | 786 | |||
AK308245.1 | - | 5 | 1067 | |||
BC004924.2 | - | 6 | 773 | 3605116 | ||
BC008832.2 | - | 6 | 806 | 4308914 | ||
BC015789.1 | - | 5 | 782 | 4866014 | ||
BC063434.1 | - | 5 | 698 | 5399570 | ||
BC067083.1 | - | 6 | 781 | 4110270 | ||
BC071863.1 | - | 4 | 386 | 4651720 | ||
BC071864.1 | - | 6 | 801 | 6083428 | ||
CX785762.1 | - | 5 | 714 | |||
M23614.1 | - | 5 | 813 | |||
M23615.1 | - | 5 | 780 | |||
M23616.1 | - | 6 | 815 | |||
M23617.1 | - | 5 | 708 | |||
M23618.1 | - | 4 | 615 | |||
M23619.1 | - | 6 | 918 | |||
X14957.1 | - | 6 | 835 | |||
X14958.1 | - | 6 | 802 |