Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HMGA1 SIRNA Assays ›
Gene Symbol:
HMGA1
Gene Name:
high mobility group AT-hook 1
Gene Aliases:
HMG-R, HMGA1A, HMGIY
Chromosome Location:
Chr.6: 34236800 - 34246231 on Build GRCh38
UniGene ID:
Hs.518805
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s6669
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HMGA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001319077.1
NP_001306006.1
4
477
NM_001319078.1
NP_001306007.1
3
398
NM_001319079.1
NP_001306008.1
4
510
NM_001319080.1
NP_001306009.1
4
509
NM_001319081.1
NP_001306010.1
3
396
NM_001319082.1
NP_001306011.1
4
532
NM_002131.3
NP_002122.1
4
479
NM_145899.2
NP_665906.1
4
512
NM_145901.2
NP_665908.1
3
679
NM_145902.2
NP_665909.1
3
646
NM_145903.2
NP_665910.1
3
365
NM_145905.2
NP_665912.1
3
368
GenBank mRNA
AK096863.1
-
5
418
AK308245.1
-
3
652
BC004924.2
-
4
359
3605116
BC008832.2
-
4
392
4308914
BC015789.1
-
3
368
4866014
BC063434.1
-
3
284
5399570
BC067083.1
-
4
367
4110270
BC071863.1
-
3
222
4651720
BC071864.1
-
4
387
6083428
BE269847.1
-
4
393
BT006774.1
-
2
157
CX785762.1
-
3
300
DA049639.1
-
4
513
DA467015.1
-
4
437
DA574958.1
-
3
347
DA936387.1
-
4
434
DB029421.1
-
4
459
KF731633.1
-
2
190
M23614.1
-
3
397
M23615.1
-
3
364
M23616.1
-
4
399
M23617.1
-
3
292
M23618.1
-
2
199
M23619.1
-
4
502
X14957.1
-
4
421
X14958.1
-
4
388
Back To Top
More Information
Panther Classification:
Molecular Function -
endodeoxyribonuclease
Gene Ontology Categories:
Function(s)
Process(es)
DNA unwinding involved in DNA replication
nucleosome disassembly
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
protein complex assembly
negative regulation of cell proliferation
response to virus
negative regulation of chromatin silencing
senescence-associated heterochromatin focus assembly
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
nuclear transport
establishment of integrated proviral latency
oncogene-induced cell senescence
positive regulation of cellular senescence
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
DNA binding
AT DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
enzyme binding
ligand-dependent nuclear receptor transcription coactivator activity
retinoic acid receptor binding
peroxisome proliferator activated receptor binding
retinoid X receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qskjf:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0