Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnaq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008139.5 | NP_032165.3 | 2, 3 | 818 | ||
GenBank mRNA | AK147393.1 | - | 2, 3 | 819 | ||
BC057583.1 | - | 2, 3 | 558 | 6408959 | ||
M55412.1 | - | 2, 3 | 359 |