Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038637.1 | NP_001033726.1 | 5 | 848 | ||
NM_001285908.1 | NP_001272837.1 | 4 | 808 | |||
NM_001285909.1 | NP_001272838.1 | 6 | 915 | |||
NM_001285910.1 | NP_001272839.1 | 3 | 955 | |||
NM_001285911.1 | NP_001272840.1 | 4 | 823 | |||
NM_010315.4 | NP_034445.1 | 4 | 996 | |||
GenBank mRNA | AK036138.1 | - | 4 | 798 | ||
AK158697.1 | - | 4 | 997 | |||
AK160396.1 | - | 4 | 726 |