Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008148.4 | NP_032174.2 | 2 | 846 | ||
GenBank mRNA | AK031272.1 | - | 2 | 845 | ||
AK172059.1 | - | 2 | 845 |