Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291025.1 | NP_001277954.1 | 2 | 708 | ||
NM_011822.4 | NP_035952.2 | 2 | 708 | |||
GenBank mRNA | AB008895.1 | - | 2 | 663 | ||
AK016852.1 | - | 2 | 655 | |||
AK033349.1 | - | 2 | 655 | |||
AK137390.1 | - | 2 | 651 | |||
AK155173.1 | - | 2 | 611 | |||
AK163680.1 | - | 2 | 684 | |||
AK169284.1 | - | 2 | 691 | |||
AK170431.1 | - | 2 | 655 | |||
AK183905.1 | - | 1 | 178 | |||
AK184963.1 | - | 1 | 47 | |||
AK194343.1 | - | 1 | 48 | |||
AK198137.1 | - | 1 | 47 | |||
BC014287.1 | - | 2 | 681 | 4193343 |