Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291025.1 | NP_001277954.1 | 7 | 1564 | ||
NM_011822.4 | NP_035952.2 | 6 | 1384 | |||
GenBank mRNA | AB008895.1 | - | 6 | 1339 | ||
AK155173.1 | - | 5 | 1565 | |||
AK163680.1 | - | 6 | 1360 | |||
AK169284.1 | - | 6 | 1367 | |||
AK170431.1 | - | 7 | 1511 | |||
AK195137.1 | - | 3 | 313 | |||
AK197337.1 | - | 2 | 164 | |||
BC003917.1 | - | 4 | 474 | |||
BC014287.1 | - | 6 | 1357 | 4193343 |