Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010338.2 | NP_034468.2 | 2 | 2602 | ||
GenBank mRNA | AJ223305.1 | - | 3355 | |||
AK032525.1 | - | 2 | 2438 | |||
AK053287.1 | - | 1154 | ||||
BC031918.1 | - | 2 | 2470 | 5364562 |
Set Membership: |
G-protein coupled receptor |