Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmur1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010341.1 | NP_034471.1 | 1 | 53 | ||
GenBank mRNA | AF044602.1 | - | 1 | 53 | ||
BC120822.1 | - | 1 | 74 | 40129745 | ||
BC137775.1 | - | 1 | 74 | 8860794 |
Set Membership: |
![]() |