Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lancl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190984.1 | NP_001177913.1 | 7 | 871 | ||
NM_001190985.1 | NP_001177914.1 | 6 | 1089 | |||
NM_021295.3 | NP_067270.1 | 7 | 905 | |||
GenBank mRNA | AJ294535.1 | - | 7 | 898 | ||
AK146120.1 | - | 6 | 1080 | |||
AK182820.1 | - | 1 | 19 | |||
BC038649.1 | - | 4 | 410 | |||
BC058560.1 | - | 7 | 871 | 6315377 | ||
Y11550.1 | - | 7 | 870 | |||
Y16518.1 | - | 6 | 1049 |