Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Grin2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010350.2 | NP_034480.2 | 8, 9 | 2259 | ||
GenBank mRNA | AK147390.1 | - | 7, 8 | 1987 | ||
AK165770.1 | - | 5, 6 | 1114 | |||
BC137849.1 | - | 6, 7 | 1649 | 8860868 | ||
D10694.1 | - | 5, 6 | 1486 |