Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008175.4 | NP_032201.2 | 8 | 985 | ||
GenBank mRNA | AK013678.1 | - | 8 | 801 | ||
AK018744.1 | - | 8 | 826 | |||
AK147932.1 | - | 8 | 802 | |||
AK149570.1 | - | 8 | 804 | |||
AK150125.1 | - | 8 | 800 | |||
AK150205.1 | - | 8 | 801 | |||
AK150338.1 | - | 8 | 790 | |||
AK150420.1 | - | 8 | 789 | |||
AK150717.1 | - | 8 | 801 | |||
AK150746.1 | - | 8 | 802 | |||
AK150881.1 | - | 8 | 802 | |||
AK151007.1 | - | 8 | 801 | |||
AK151162.1 | - | 8 | 802 | |||
AK151343.1 | - | 8 | 801 | |||
AK151713.1 | - | 8 | 801 | |||
AK151756.1 | - | 8 | 801 | |||
AK152048.1 | - | 8 | 801 | |||
AK152087.1 | - | 8 | 801 | |||
AK152118.1 | - | 8 | 790 | |||
AK152213.1 | - | 8 | 790 | |||
AK152705.1 | - | 8 | 801 | |||
AK152765.1 | - | 8 | 801 | |||
AK152944.1 | - | 8 | 790 | |||
AK153183.1 | - | 8 | 790 | |||
AK153269.1 | - | 8 | 801 | |||
AK153289.1 | - | 8 | 801 | |||
AK153418.1 | - | 8 | 801 | |||
AK153470.1 | - | 8 | 801 | |||
AK154492.1 | - | 8 | 801 | |||
AK159397.1 | - | 8 | 790 | |||
AK159428.1 | - | 8 | 790 | |||
AK159557.1 | - | 8 | 801 | |||
AK159809.1 | - | 8 | 801 | |||
AK160022.1 | - | 8 | 790 | |||
AK160033.1 | - | 8 | 801 | |||
AK160074.1 | - | 8 | 790 | |||
AK179088.1 | - | 3 | 297 | |||
AK182648.1 | - | 286 | ||||
AK193719.1 | - | 3 | 284 | |||
AK211032.1 | - | 3 | 254 | |||
AK219696.1 | - | 3 | 264 | |||
BC037047.1 | - | 7 | 782 | |||
BC049943.1 | - | 8 | 787 | |||
BC129849.1 | - | 8 | 785 | 40109630 | ||
BC129850.1 | - | 8 | 785 | 40109631 | ||
X62321.1 | - | 7 | 766 |