Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010358.5 | NP_034488.1 | 2, 3 | 318 | ||
GenBank mRNA | BC003822.1 | - | 2, 3 | 141 | 3600444 | |
BC046758.1 | - | 2, 3 | 171 | 5693552 | ||
BC091763.1 | - | 2, 3 | 151 | 5102093 | ||
J03952.1 | - | 2, 3 | 132 | |||
J04632.1 | - | 2, 3 | 319 |
Gene Symbol: Gstm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC019413.1 | - | 2, 3 | 151 | 4166881 |