Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Guca2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008191.2 | NP_032217.2 | 3 | 467 | ||
GenBank mRNA | AK002364.1 | - | 3 | 439 | ||
AV002130.1 | - | 2 | 220 | |||
BC024373.1 | - | 3 | 434 | 4971289 | ||
CF423670.1 | - | 3 | 480 | |||
U67800.1 | - | 3 | 423 | |||
U90727.1 | - | 3 | 469 |