Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029850.4 | NP_001025021.1 | 23 | 4130 | ||
NM_001083320.2 | NP_001076789.1 | 22 | 3505 | |||
NM_001083321.2 | NP_001076790.1 | 22 | 3505 | |||
NM_001286784.1 | NP_001273713.1 | 22 | 4127 | |||
NM_001286785.1 | NP_001273714.1 | 23 | 4166 | |||
NM_001286786.1 | NP_001273715.1 | 22 | 4127 | |||
NM_001286788.1 | NP_001273717.1 | 22 | 3499 | |||
NM_010367.3 | NP_034497.1 | 20 | 3839 | |||
GenBank mRNA | AB194411.1 | - | 22 | 3532 | ||
AB194412.1 | - | 22 | 3532 | |||
AF027503.1 | - | 20 | 4168 | |||
AK032350.1 | - | 10 | 1221 | |||
AK147358.1 | - | 23 | 4167 | |||
AK147666.1 | - | 20 | 3068 | |||
AK163582.1 | - | 16 | 2689 | |||
AK163800.1 | - | 22 | 3370 | |||
AK220499.1 | - | 22 | 3204 | |||
AY497557.1 | - | 23 | 3535 | |||
BC060273.1 | - | 22 | 3505 | |||
BC095943.1 | - | 21 | 3853 | |||
BC150820.1 | - | 23 | 3840 | 9087731 | ||
CF726496.1 | - | 2 | 37 |