Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029850.4 | NP_001025021.1 | 14 | 2976 | ||
NM_001083320.2 | NP_001076789.1 | 13 | 2315 | |||
NM_001083321.2 | NP_001076790.1 | 13 | 2318 | |||
NM_001286784.1 | NP_001273713.1 | 13 | 2940 | |||
NM_001286785.1 | NP_001273714.1 | 14 | 2976 | |||
NM_001286786.1 | NP_001273715.1 | 13 | 2940 | |||
NM_001286788.1 | NP_001273717.1 | 13 | 2318 | |||
NM_010367.3 | NP_034497.1 | 13 | 2937 | |||
GenBank mRNA | AB194411.1 | - | 13 | 2345 | ||
AB194412.1 | - | 13 | 2345 | |||
AF027503.1 | - | 13 | 3266 | |||
AK032350.1 | - | 1 | 31 | |||
AK147358.1 | - | 14 | 2977 | |||
AK147666.1 | - | 12 | 1998 | |||
AK163582.1 | - | 10 | 1643 | |||
AK163800.1 | - | 13 | 2183 | |||
AK220499.1 | - | 13 | 2023 | |||
AY497557.1 | - | 14 | 2381 | |||
BB653276.1 | - | 2 | 383 | |||
BC060273.1 | - | 13 | 2315 | |||
BC095943.1 | - | 14 | 2951 | |||
BC150820.1 | - | 14 | 2650 | 9087731 |