Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029850.4 | NP_001025021.1 | 5 | 1458 | ||
NM_001083320.2 | NP_001076789.1 | 4 | 800 | |||
NM_001083321.2 | NP_001076790.1 | 4 | 800 | |||
NM_001286784.1 | NP_001273713.1 | 5 | 1458 | |||
NM_001286785.1 | NP_001273714.1 | 5 | 1458 | |||
NM_001286786.1 | NP_001273715.1 | 5 | 1458 | |||
NM_001286788.1 | NP_001273717.1 | 4 | 800 | |||
NM_010367.3 | NP_034497.1 | 5 | 1458 | |||
GenBank mRNA | AB194411.1 | - | 5 | 863 | ||
AB194412.1 | - | 5 | 863 | |||
AF027503.1 | - | 5 | 1787 | |||
AK147358.1 | - | 5 | 1459 | |||
AK147666.1 | - | 4 | 515 | |||
AK163582.1 | - | 2 | 161 | |||
AK163800.1 | - | 4 | 665 | |||
AK220499.1 | - | 4 | 505 | |||
AY497557.1 | - | 5 | 863 | |||
BC060273.1 | - | 4 | 800 | |||
BC095943.1 | - | 5 | 1436 | |||
BC150820.1 | - | 5 | 1132 | 9087731 |