Search Thermo Fisher Scientific
Gene Symbol: Gys1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030678.3 | NP_109603.2 | 4, 5 | 891 | ||
GenBank mRNA | AK050813.1 | - | 2, 3 | 324 | ||
AK169344.1 | - | 4, 5 | 887 | |||
AK170404.1 | - | 4, 5 | 890 | |||
AK170585.1 | - | 4, 5 | 811 | |||
AK171148.1 | - | 4, 5 | 846 | |||
AK171426.1 | - | 4, 5 | 870 | |||
AK205458.1 | - | 1, 2 | 88 | |||
AK207597.1 | - | 1, 2 | 88 | |||
BC019389.1 | - | 4, 5 | 684 | 3663506 | ||
BC131687.1 | - | 4, 5 | 738 | 40111722 | ||
BC131688.1 | - | 4, 5 | 738 | 40111723 | ||
BC152550.1 | - | 4, 5 | 738 | 40111724 | ||
X94616.1 | - | 4, 5 | 738 |