Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gzmf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010374.3 | NP_034504.1 | 5 | 699 | ||
GenBank mRNA | BC100311.1 | - | 5 | 699 | 30523018 | |
BC145435.1 | - | 5 | 675 | 9053993 | ||
J03257.1 | - | 5 | 709 | |||
X12823.1 | - | 5 | 650 |