Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pfdn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185182.1 | NP_001172111.1 | 4 | 215 | ||
NM_010385.2 | NP_034515.1 | 3 | 419 | |||
GenBank mRNA | AA023721.1 | - | 3 | 195 | ||
AK010679.1 | - | 3 | 409 | |||
AK076015.1 | - | 3 | 402 | |||
AK167900.1 | - | 3 | 405 | |||
AV446044.1 | - | 2 | 74 | |||
BC022974.1 | - | 3 | 385 | 5373335 | ||
KJ650212.1 | - | 3 | 163 | |||
M65255.1 | - | 3 | 382 |