Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hadh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008212.4 | NP_032238.2 | 8 | 971 | ||
GenBank mRNA | AK075609.1 | - | 8 | 897 | ||
AK132260.1 | - | 8 | 934 | |||
AK148486.1 | - | 8 | 908 | |||
AK167024.1 | - | 8 | 898 | |||
AK168238.1 | - | 8 | 918 | |||
AK168877.1 | - | 8 | 898 | |||
AK169238.1 | - | 6 | 527 | |||
AK169261.1 | - | 8 | 937 | |||
AK208263.1 | - | 5 | 349 | |||
BC028833.1 | - | 8 | 803 | |||
BC064712.1 | - | 8 | 913 | 30303218 | ||
D29639.1 | - | 8 | 897 |