Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008214.4 | NP_032240.3 | 4 | 448 | ||
GenBank mRNA | AK011685.1 | - | 4 | 364 | ||
AK077437.1 | - | 4 | 364 | |||
AK088824.1 | - | 4 | 401 | |||
AK146922.1 | - | 4 | 401 | |||
AK152329.1 | - | 4 | 380 | |||
AK166826.1 | - | 4 | 364 | |||
AK166892.1 | - | 4 | 411 | |||
AK211613.1 | - | 3 | 264 | |||
BC020088.1 | - | 4 | 383 | 4008479 | ||
CA561201.1 | - | 4 | 448 | |||
U39473.1 | - | 4 | 428 |